Learn More
Medchemexpress LLC Rho-kinase-IN-1 | 1035094-83-7 | 99.9% | 352.50 g/mol | C20H24N4S | 100 MG
SDP
Catalog No. 5000214516
Encompass_Preferred
Rho-Kinase-IN-1 is a small-molecule Rho-associated kinase (ROCK) inhibitor used in biochemical and cellular research. It demonstrates potent activity against ROCK isoforms (Ki = 30.5 nM for ROCK1; Ki = 3.9 nM for ROCK2) and is employed in studies of cell proliferation, tissue remodeling, vascular permeability, and inflammatory processes. Analytical documentation is available from the supplier.
Catalog No. 50-002-14516 Supplier Medchemexpress LLC Supplier No. HY100270100MG
Rho-Kinase-IN-1 is a small-molecule Rho-associated kinase (ROCK) inhibitor used in biochemical and cellular research. It demonstrates potent activity against ROCK isoforms (Ki = 30.5 nM for ROCK1; Ki = 3.9 nM for ROCK2) and is employed in studies of cell proliferation, tissue remodeling, vascular permeability, and inflammatory processes. Analytical documentation is available from the supplier.