Learn More
Medchemexpress LLC Rho-kinase-IN-1 | 1035094-83-7 | MFCD18633216 | 99.9% | 352.50 g·mol⁻¹ | C20H24N4S | 1 MG
SDP
Catalog No. 5000214322
Encompass_Preferred
Rho-Kinase-IN-1 is a small-molecule inhibitor of Rho-associated kinases used in biochemical and cellular research. It shows high potency against ROCK isoforms and is provided as a high-purity solid for assay development, screening, and mechanistic studies.
  • Selective inhibition of ROCK1 and ROCK2 with Ki = 30.5 nM and 3.9 nM.
  • High purity for reliable biochemical and cell-based assays.
  • Supplied as a small solid quantity suitable for screening and dose-response experiments.
  • Chemical formula C20H24N4S; molecular weight 352.50 g·mol⁻¹.
  • Useful in studies of cytoskeletal regulation, cell motility, and related signaling pathways.
Catalog No. 50-002-14322 Supplier Medchemexpress LLC Supplier No. HY1002701MG
Rho-Kinase-IN-1 is a small-molecule inhibitor of Rho-associated kinases used in biochemical and cellular research. It shows high potency against ROCK isoforms and is provided as a high-purity solid for assay development, screening, and mechanistic studies.
  • Selective inhibition of ROCK1 and ROCK2 with Ki = 30.5 nM and 3.9 nM.
  • High purity for reliable biochemical and cell-based assays.
  • Supplied as a small solid quantity suitable for screening and dose-response experiments.
  • Chemical formula C20H24N4S; molecular weight 352.50 g·mol⁻¹.
  • Useful in studies of cytoskeletal regulation, cell motility, and related signaling pathways.