Learn More
Medchemexpress LLC Rho-kinase-IN-1 | 1035094-83-7 | MFCD18633216 | 99.9% | 352.50 g·mol⁻¹ | C20H24N4S | 25 MG
SDP
Catalog No. 5000214535
Encompass_Preferred
Rho-Kinase-IN-1 is a selective Rho-associated protein kinase (ROCK) inhibitor used for laboratory research. Reported in patent US20090325960A1 (compound 1.008), it exhibits potent activity against ROCK1 and ROCK2 and is used to study signaling pathways involved in cell proliferation, cytoskeletal remodeling, edema, and inflammation. For research use only.
  • Potent inhibition of ROCK1 and ROCK2 (Ki ≈ 30.5 nM and 3.9 nM).
  • High purity suitable for biochemical and cellular assays (99.91%).
  • Molecular formula C20H24N4S and molecular weight 352.50 g·mol⁻¹.
  • Available as solid and as DMSO solution for assay flexibility.
  • Supplied with datasheet and SDS for safety and handling information.
Catalog No. 50-002-14535 Supplier Medchemexpress LLC Supplier No. HY10027025MG
Rho-Kinase-IN-1 is a selective Rho-associated protein kinase (ROCK) inhibitor used for laboratory research. Reported in patent US20090325960A1 (compound 1.008), it exhibits potent activity against ROCK1 and ROCK2 and is used to study signaling pathways involved in cell proliferation, cytoskeletal remodeling, edema, and inflammation. For research use only.
  • Potent inhibition of ROCK1 and ROCK2 (Ki ≈ 30.5 nM and 3.9 nM).
  • High purity suitable for biochemical and cellular assays (99.91%).
  • Molecular formula C20H24N4S and molecular weight 352.50 g·mol⁻¹.
  • Available as solid and as DMSO solution for assay flexibility.
  • Supplied with datasheet and SDS for safety and handling information.