Learn More
Medchemexpress LLC Rho-kinase-in-1 | 1035094-83-7 | MFCD18633216 | 99.9% | 352.50 g·mol⁻¹ | C20H24N4S | 50 MG
SDP
Catalog No. 5000214451
Encompass_Preferred
Rho-Kinase-IN-1 is a selective Rho-associated protein kinase (ROCK) inhibitor used in preclinical and in vitro research. It inhibits ROCK1 and ROCK2 with reported Ki values of 30.5 nM and 3.9 nM, respectively, and is cited from patent US20090325960A1. The compound is supplied as a high-purity research reagent for studies of cell proliferation, cytoskeletal remodeling, edema, and inflammatory processes.
  • Potent inhibition of ROCK1 and ROCK2 (Ki 30.5 nM and 3.9 nM)
  • High purity suitable for research (99.91%)
  • Molecular formula C20H24N4S; molecular weight 352.50 g·mol⁻¹
  • Suitable for in vitro and preclinical studies of cell proliferation and remodeling
  • Available in multiple pack sizes for laboratory use
Catalog No. 50-002-14451 Supplier Medchemexpress LLC Supplier No. HY10027050MG
Rho-Kinase-IN-1 is a selective Rho-associated protein kinase (ROCK) inhibitor used in preclinical and in vitro research. It inhibits ROCK1 and ROCK2 with reported Ki values of 30.5 nM and 3.9 nM, respectively, and is cited from patent US20090325960A1. The compound is supplied as a high-purity research reagent for studies of cell proliferation, cytoskeletal remodeling, edema, and inflammatory processes.
  • Potent inhibition of ROCK1 and ROCK2 (Ki 30.5 nM and 3.9 nM)
  • High purity suitable for research (99.91%)
  • Molecular formula C20H24N4S; molecular weight 352.50 g·mol⁻¹
  • Suitable for in vitro and preclinical studies of cell proliferation and remodeling
  • Available in multiple pack sizes for laboratory use