Learn More
Medchemexpress LLC HY-100270 5mg Medchemexpress, Rho-Kinase-IN-1 CAS:1035094-83-7 Purity:>98%
SDP
Catalog No. 501967720
Encompass_Preferred
Medchemexpress, HY-100270 5mg Rho-Kinase-IN-1 CAS:1035094-83-7 Rho-Kinase-IN-1 is a Rho kinase (ROCK) inhibitor (Ki values of 30.5 and 3.9 nM for ROCK1 and ROCK2, respectively) extracted from US20090325960A1, compound 1.008. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7720 Supplier Medchemexpress LLC Supplier No. HY1002705MG
Medchemexpress, HY-100270 5mg Rho-Kinase-IN-1 CAS:1035094-83-7 Rho-Kinase-IN-1 is a Rho kinase (ROCK) inhibitor (Ki values of 30.5 and 3.9 nM for ROCK1 and ROCK2, respectively) extracted from US20090325960A1, compound 1.008. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.