Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rho-Kinase-IN-2 | 2573071-18-6 | C20H25FN4O2 | 25 MG
SDP

Catalog No. 5000374442
Encompass_Preferred

Rho-Kinase-IN-2 (Compound 23) is an orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor with an IC50 of 3 nM for ROCK2. It can be used in Huntington's research. This product is for research use only and not sold to patients. It appears as a solid, light yellow to yellow in color, with a molecular weight of 372.44.

  • Orally active
  • Selective and CNS-penetrant Rho Kinase (ROCK) inhibitor
  • ROCK2 IC50 of 3 nM
  • Useful in Huntington's research
  • Solid, light yellow to yellow appearance
  • Molecular weight of 372.44
  • CAS number: 2573071-18-6
  • Chemical formula: C20H25FN4O2

Catalog No. 50-003-74442 Supplier Medchemexpress LLC Supplier No. 25MGHY150640
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rho-Kinase-IN-2 (Compound 23) is an orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor with an IC50 of 3 nM for ROCK2. It can be used in Huntington's research. This product is for research use only and not sold to patients. It appears as a solid, light yellow to yellow in color, with a molecular weight of 372.44.

  • Orally active
  • Selective and CNS-penetrant Rho Kinase (ROCK) inhibitor
  • ROCK2 IC50 of 3 nM
  • Useful in Huntington's research
  • Solid, light yellow to yellow appearance
  • Molecular weight of 372.44
  • CAS number: 2573071-18-6
  • Chemical formula: C20H25FN4O2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.