Learn More
Medchemexpress LLC Rho-Kinase-IN-2 | 2573071-18-6 | C20H25FN4O2 | 50 MG
SDP
Catalog No. 5000375179
Encompass_Preferred
Rho-Kinase-IN-2 (Compound 23) is an orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor (ROCK2 IC50 = 3 nM). It can be used in Huntington's research. In vitro studies show it increases AKT phosphorylation and decreases MYPT1 phosphorylation. In vivo, it demonstrates dose- and time-dependent ROCK1 and ROCK2 target engagement and tolerability in mouse models. Oral administration decreases mean arterial, systolic, and diastolic blood pressure.
  • Orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor
  • ROCK2 IC50 of 3 nM
  • Increases AKT phosphorylation and decreases MYPT1 phosphorylation in A7r5 and PANC1 cells
  • Demonstrates dose- and time-dependent ROCK1 and ROCK2 target engagement in mice
  • Used in Huntington's research
Catalog No. 50-003-75179 Supplier Medchemexpress LLC Supplier No. HY15064050MG
Rho-Kinase-IN-2 (Compound 23) is an orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor (ROCK2 IC50 = 3 nM). It can be used in Huntington's research. In vitro studies show it increases AKT phosphorylation and decreases MYPT1 phosphorylation. In vivo, it demonstrates dose- and time-dependent ROCK1 and ROCK2 target engagement and tolerability in mouse models. Oral administration decreases mean arterial, systolic, and diastolic blood pressure.
  • Orally active, selective, and CNS-penetrant Rho Kinase (ROCK) inhibitor
  • ROCK2 IC50 of 3 nM
  • Increases AKT phosphorylation and decreases MYPT1 phosphorylation in A7r5 and PANC1 cells
  • Demonstrates dose- and time-dependent ROCK1 and ROCK2 target engagement in mice
  • Used in Huntington's research