Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP8868EC10UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: E. coli. Species Reactivity: E. coli. Protein Expression: A DNA sequence encoding the Escherichia coli (strain K12) Transcription termination factor Rho, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: rho. Gene ID: 948297. Uniprot: P0AG30.
Catalog No. 50-188-8204