Learn More
Cytoskeleton, Inc. RhoA G-LISA Activation Assay Kit (Colorimetric Format), 24 Assays
Catalog No. NC0983585
Encompass
The kit contains sufficient reagents to perform 24 RhoA activation assays. Since the Rho-GTP affinity wells are supplied as strips and the strips can be broken into smaller pieces, each kit can be used for anywhere from one to multiple assays.
Catalog No. NC0983585 Supplier Cytoskeleton, Inc. Supplier No. BK124S
The kit contains sufficient reagents to perform 24 RhoA activation assays. Since the Rho-GTP affinity wells are supplied as strips and the strips can be broken into smaller pieces, each kit can be used for anywhere from one to multiple assays.