Learn More
NEW EAST BIOSCIENCES LLC RhoA Pull-Down Activation Assay Kit, 30 Assays
Catalog No. NC2750676
Encompass_Preferred
The RhoA Activation Assay Kit uses configuration-specific anti-RhoA-GTP Mouse monoclonal antibody to measure RhoA-GTP levels in cell extracts or in vitro GTPγS loading RhoA activation assays. Anti-RhoA-GTP mouse monoclonal antibody is first incubated with cell lysates containing RhoA-GTP. Next, the GTP-bound RhoA is pulled down by protein A/G agarose. Finally, the precipitated RhoA-GTP is detected through immunoblot analysis using Anti-RhoA Rabbit Polyclonal Antibody.
Catalog No. NC2750676 Supplier NEW EAST BIOSCIENCES LLC Supplier No. 80601
Add to Cart
Add to Cart
The RhoA Activation Assay Kit uses configuration-specific anti-RhoA-GTP Mouse monoclonal antibody to measure RhoA-GTP levels in cell extracts or in vitro GTPγS loading RhoA activation assays. Anti-RhoA-GTP mouse monoclonal antibody is first incubated with cell lysates containing RhoA-GTP. Next, the GTP-bound RhoA is pulled down by protein A/G agarose. Finally, the precipitated RhoA-GTP is detected through immunoblot analysis using Anti-RhoA Rabbit Polyclonal Antibody.