Learn More
Aviva Systems Biology Corp RHOB Antibody - middle region (ARP54702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RHOB. Alternate names: ARH6, ARHB, MST081, MSTP081, RHOH6ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62987