Learn More
Aviva Systems Biology Corp RHOBTB1 Antibody - middle region (ARP41883_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41883_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOBTB1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RHOBTB1. Alternate names: KIAA0740, MGC33059, MGC33841ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53317