Learn More
Aviva Systems Biology Corp RHOC Antibody - N-terminal region (ARP55742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RHOC. Alternate names: ARH9, ARHC, H9, MGC1448, MGC61427, RHOH9ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-61594