Learn More
Medchemexpress LLC HY-D0989 1mg Medchemexpress, Rhod-2 AM CAS:145037-81-6 Purity:>98%
SDP
Catalog No. 501970193
Encompass_Preferred
Medchemexpress, HY-D0989 1mg Rhod-2 AM CAS:145037-81-6 Rhod-2 AM is a fluorescent, mitochondrial probe (λex=552 nm, λem=581 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0193 Supplier Medchemexpress LLC Supplier No. HYD09891MG
Medchemexpress, HY-D0989 1mg Rhod-2 AM CAS:145037-81-6 Rhod-2 AM is a fluorescent, mitochondrial probe (λex=552 nm, λem=581 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.