Learn More
Biotium Rhod-2, AM Ester
Catalog No. NC0322564
Encompass_Preferred
Membrane-permeant form of Rhod-2 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 is often used as a dispersing agent to facilitate dye loading. Rhod-2/AM itself does not bind Ca2 , but it is readily hydrolyzed to Rhod-2 by endogenous esterases once it enters cells. Features: Red solid soluble in DMSO; Store at -20°C and protect from light, especially in solution; C52H59BrN4O19; MW: 1132; [145037-81-6]
Catalog No. NC0322564 Supplier Biotium Supplier No. 50024
Membrane-permeant form of Rhod-2 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 is often used as a dispersing agent to facilitate dye loading. Rhod-2/AM itself does not bind Ca2 , but it is readily hydrolyzed to Rhod-2 by endogenous esterases once it enters cells. Features: Red solid soluble in DMSO; Store at -20°C and protect from light, especially in solution; C52H59BrN4O19; MW: 1132; [145037-81-6]