Learn More
Enzo Life Sciences Rhod-2 AM (ultra pure) (1mg). CAS: 129787-64-0
SDP
Catalog No. 502012608
Encompass_Preferred
Rhod-2 AM has the longest fluorescent emission signal of the commonly used calcium indicators. It contains a rhodamine-like fluorophore, making it suitable for use with argon and krypton laser excitation sources. Rhod-2 signal intensity one of the strongest of all the calcium probes. Rhod 2 is especially suitable probe for intracellular calcium monitoring by confocal laser-scanning microscopy and flow cytometry. The dissociation constant of Rhod 2 with calcium is among the highest of all the fluorescent calcium probes, providing a wider range for monitoring calcium concentration. Wavelength Maxima: Excitation 549nm, Emission 578nm. Alternative name: 1-[2-Amino-5-(3-dimethylamino-6-dimethylammonio-9-xanthenyl)phenoxy]-2-(2-amino-5-methylphenoxy)]ethane-N,N,N',N'-tetraacetic acid, tetraacetoxy methyl ester . Br. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2608 Supplier Enzo Life Sciences Supplier No. ENZ52010
Rhod-2 AM has the longest fluorescent emission signal of the commonly used calcium indicators. It contains a rhodamine-like fluorophore, making it suitable for use with argon and krypton laser excitation sources. Rhod-2 signal intensity one of the strongest of all the calcium probes. Rhod 2 is especially suitable probe for intracellular calcium monitoring by confocal laser-scanning microscopy and flow cytometry. The dissociation constant of Rhod 2 with calcium is among the highest of all the fluorescent calcium probes, providing a wider range for monitoring calcium concentration. Wavelength Maxima: Excitation 549nm, Emission 578nm. Alternative name: 1-[2-Amino-5-(3-dimethylamino-6-dimethylammonio-9-xanthenyl)phenoxy]-2-(2-amino-5-methylphenoxy)]ethane-N,N,N',N'-tetraacetic acid, tetraacetoxy methyl ester . Br. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.