Learn More
Biotium Rhod-2, Tripotassium Salt
Catalog No. 501964689
Encompass_Preferred
Membrane-impermeant calcium indictor that can be loaded into cells via microinjection or scrape loading. Properties: Kd: 1 uM; Ex/Em (low or high [Ca2 ])= 556nm/576nm; e (556 nm) = 80,000 M-1cm-1; Orange red solid soluble in DMSO and water (pH >6); Store at 4°C and protect from light, especially in solution; C40H39K3N4O11; MW: 869
Catalog No. 50-196-4689 Supplier Biotium Supplier No. 50021
Membrane-impermeant calcium indictor that can be loaded into cells via microinjection or scrape loading. Properties: Kd: 1 uM; Ex/Em (low or high [Ca2 ])= 556nm/576nm; e (556 nm) = 80,000 M-1cm-1; Orange red solid soluble in DMSO and water (pH >6); Store at 4°C and protect from light, especially in solution; C40H39K3N4O11; MW: 869