Learn More
Aviva Systems Biology Corp RHOD Antibody - C-terminal region (ARP42414_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42414_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RHOD. Alternate names: ARHD, RHOHP1, RHOM, RhoApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58218