Learn More
Abcam Rhodadyn™ B10, Negative control for Rhodadyn™ C10, 5MG
Catalog No. 502707256
Encompass_Preferred
MW 322.44 g/mol, Purity >98%. Negative control for Rhodadyn™ C10 (ab120795). Demonstrates no significant inhibition of dynamin I or dynamin II in vitro GTPase activity at concentrations up to 300 μM and has no effect on clathrin-mediated endocytosis of transferrin when applied for 30 minutes to U2OS cells in culture. Robertson J et al. The rhodadyn™ a new class of small molecule inhibitors of dynamin GTPase. ACS Med. Chem (2012).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7256 Supplier Abcam Supplier No. AB1207965MG
MW 322.44 g/mol, Purity >98%. Negative control for Rhodadyn™ C10 (ab120795). Demonstrates no significant inhibition of dynamin I or dynamin II in vitro GTPase activity at concentrations up to 300 μM and has no effect on clathrin-mediated endocytosis of transferrin when applied for 30 minutes to U2OS cells in culture. Robertson J et al. The rhodadyn™ a new class of small molecule inhibitors of dynamin GTPase. ACS Med. Chem (2012).
The product is subject to the following: Abcam Restricted Use Statement