Learn More
Cayman Chemical Rhodamine 101 (inner salt), Purity: ≥95%, CAS: 116450-56-7, 100 mg
SDP
Catalog No. 502983624
Encompass_Preferred
A bright fluorescent dye with excitation and emission maxima of 565 and 595 nm, respectively
Catalog No. 50-298-3624 Supplier Cayman Chemical Supplier No. 20654100 MG
A bright fluorescent dye with excitation and emission maxima of 565 and 595 nm, respectively