Learn More
Cayman Chemical Rhodamine 110 (chloride), Purity: ≥97%, CAS: 13558-31-1, 1 g
SDP
Catalog No. 502984587
Encompass_Preferred
A green fluorescent cationic dye (ex/em max 496/520 nm) that is incorporated with a hydrolytic substrate (e.g., proteinase or peptidase substrates) for use as a highly sensitive detection reagent in fluorescence-based enzyme assays
Catalog No. 50-298-4587 Supplier Cayman Chemical Supplier No. 190611 G
A green fluorescent cationic dye (ex/em max 496/520 nm) that is incorporated with a hydrolytic substrate (e.g., proteinase or peptidase substrates) for use as a highly sensitive detection reagent in fluorescence-based enzyme assays