Learn More
Cayman Chemical Rhodamine 110 (chloride), Purity: ≥97%, CAS: 13558-31-1, 250 mg
SDP
Catalog No. 502983775
Encompass_Preferred
A green fluorescent cationic dye (ex/em max 496/520 nm) that is incorporated with a hydrolytic substrate (e.g., proteinase or peptidase substrates) for use as a highly sensitive detection reagent in fluorescence-based enzyme assays
Catalog No. 50-298-3775 Supplier Cayman Chemical Supplier No. 19061250 MG
A green fluorescent cationic dye (ex/em max 496/520 nm) that is incorporated with a hydrolytic substrate (e.g., proteinase or peptidase substrates) for use as a highly sensitive detection reagent in fluorescence-based enzyme assays