Learn More
Cayman Chemical Rhodamine 123 (chloride), Purity: ≥95%, CAS: 62669-70-9, 100 mg
SDP
Catalog No. 502965217
Encompass_Preferred
A membrane-permeable cationic dye that is a substrate for P-gp and is rapidly exported from cells with functional P-gp; also accumulates within mitochondria and can inhibit oxidative phosphorylation; ex/em = 507/529 nm
Catalog No. 50-296-5217 Supplier Cayman Chemical Supplier No. 16672100 MG
A membrane-permeable cationic dye that is a substrate for P-gp and is rapidly exported from cells with functional P-gp; also accumulates within mitochondria and can inhibit oxidative phosphorylation; ex/em = 507/529 nm