Learn More
Cayman Chemical RhodamIn B hydrazIde 100mg
SDP
Catalog No. 502994010
Encompass_Preferred
A water-soluble fluorescent probe; ex/em spectra = 510/578 nm, respectively; has been used for the detection of copper, peroxynitrite, nitric oxide, hydrogen peroxide, glucose, diacetyl, and hemoglobin
Catalog No. 50-299-4010 Supplier Cayman Chemical Supplier No. 23133100 MG
A water-soluble fluorescent probe; ex/em spectra = 510/578 nm, respectively; has been used for the detection of copper, peroxynitrite, nitric oxide, hydrogen peroxide, glucose, diacetyl, and hemoglobin