Learn More
Cayman Chemical Rhodamine B hydrazide, Purity: ≥98%, CAS: 74317-53-6, 5 mg
SDP
Catalog No. 502995563
Encompass_Preferred
A water-soluble fluorescent probe; ex/em spectra = 510/578 nm, respectively; has been used for the detection of copper, peroxynitrite, nitric oxide, hydrogen peroxide, glucose, diacetyl, and hemoglobin
Catalog No. 50-299-5563 Supplier Cayman Chemical Supplier No. 231335 MG
A water-soluble fluorescent probe; ex/em spectra = 510/578 nm, respectively; has been used for the detection of copper, peroxynitrite, nitric oxide, hydrogen peroxide, glucose, diacetyl, and hemoglobin