Learn More
Cayman Chemical Rhodamine B isothiocyanate (mixed isomers), Purity: Mixture of isomers, CAS: 944130-99-8, 250 mg
SDP
Catalog No. 502984499
Encompass_Preferred
A cell-permeant fluorogenic dye used as a conjugate to antibodies and proteins for fluorescence detection in microscopy; ex/em max = 570/595 nm; used as an anterograde and retrograde tracer; used in nanoparticles to detect apoptosis
Catalog No. 50-298-4499 Supplier Cayman Chemical Supplier No. 20653250 MG
A cell-permeant fluorogenic dye used as a conjugate to antibodies and proteins for fluorescence detection in microscopy; ex/em max = 570/595 nm; used as an anterograde and retrograde tracer; used in nanoparticles to detect apoptosis