Learn More
Medchemexpress LLC HY-D1044 10mg Medchemexpress, Rhodamine 101 (chloride) CAS:64339-18-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD104410MG
Encompass_Preferred
Medchemexpress, HY-D1044 10mg Rhodamine 101 (chloride) CAS:64339-18-0 Rhodamine 101 chloride (Rhodamine 640 chloride) is a bright fluorescent dye with excitation and emission maxima of 565 and 595 nm, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0196