Learn More
Medchemexpress LLC Rhodamine 101 chloride | 64339-18-0 | 527.05 g/mol | C32H31ClN2O3 | 10 MG
SDP
Catalog No. 5000297512
Encompass_Preferred
Rhodamine 101 chloride is a bright xanthene-class fluorescent dye with excitation and emission maxima at 565 nm and 595 nm, respectively. It is used for fluorescence microscopy, cellular staining, and as a red-spectrum reference standard. The solid is supplied in small laboratory pack sizes and should be stored protected from light and moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Excitation 565 nm and emission 595 nm.
  • Suitable for fluorescence microscopy and cellular staining.
  • Useful as a reference standard in red-spectrum labeling.
  • Available in small lab pack sizes, including 10 mg.
  • Store sealed, protected from moisture and light.
Catalog No. 50-002-97512 Supplier Medchemexpress LLC Supplier No. HYD1044A10MG
Rhodamine 101 chloride is a bright xanthene-class fluorescent dye with excitation and emission maxima at 565 nm and 595 nm, respectively. It is used for fluorescence microscopy, cellular staining, and as a red-spectrum reference standard. The solid is supplied in small laboratory pack sizes and should be stored protected from light and moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Excitation 565 nm and emission 595 nm.
  • Suitable for fluorescence microscopy and cellular staining.
  • Useful as a reference standard in red-spectrum labeling.
  • Available in small lab pack sizes, including 10 mg.
  • Store sealed, protected from moisture and light.