Learn More
Medchemexpress LLC Rhodamine 101 hydrochloride | 64339-18-0 | 95.0% | 563.51 | 25 MG
SDP
Catalog No. 5000429517
Encompass_Preferred
Rhodamine 101 hydrochloride is a fluorescent dye. It is known for its bright fluorescence and specific excitation and emission characteristics.
  • Exhibits bright fluorescence
  • Excitation maxima at 565 nm
  • Emission maxima at 595 nm.
Catalog No. 50-004-29517 Supplier Medchemexpress LLC Supplier No. 25MGHYD1044A
Rhodamine 101 hydrochloride is a fluorescent dye. It is known for its bright fluorescence and specific excitation and emission characteristics.
  • Exhibits bright fluorescence
  • Excitation maxima at 565 nm
  • Emission maxima at 595 nm.