Learn More
Medchemexpress LLC Rhodamine 101 hydrochloride | 64339-18-0 | MFCD00210089 | >=95% | 563.51 | 5 MG
SDP
Catalog No. 5000428264
Encompass_Preferred
Rhodamine 101 chloride (Rhodamine 640 chloride) hydrochloride is a bright fluorescent dye with excitation and emission maxima of 565 and 595 nm, respectively.
  • For research use only.
  • We do not sell to patients.
Catalog No. 50-004-28264 Supplier Medchemexpress LLC Supplier No. HYD1044A5MG
Rhodamine 101 chloride (Rhodamine 640 chloride) hydrochloride is a bright fluorescent dye with excitation and emission maxima of 565 and 595 nm, respectively.
  • For research use only.
  • We do not sell to patients.