Learn More
Aat Bioquest Rhodamine 101 Inner Salt [CAS 116450-56-7]
SDP
Catalog No. 502979309
Encompass_Preferred
Rhodamine dyes are used extensively in a variety of biological applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.
Catalog No. 50-297-9309 Supplier Aat Bioquest Supplier No. 74
Rhodamine dyes are used extensively in a variety of biological applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.