Learn More
Abcam Rhodamine 110, 25MG
Catalog No. 50996241
Encompass_Preferred
MW 366.8 Da, Purity >98%. Green fluorophore standard widely used as the base dye in enzyme assay systems. Reference for many proteinase and peptidase substrates derived by adding amino acids or peptides to the dye's two amine functionalities.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-241 Supplier Abcam Supplier No. AB27530525MG
MW 366.8 Da, Purity >98%. Green fluorophore standard widely used as the base dye in enzyme assay systems. Reference for many proteinase and peptidase substrates derived by adding amino acids or peptides to the dye's two amine functionalities.
The product is subject to the following: Abcam Restricted Use Statement