Learn More
Medchemexpress LLC HY-D0817 50mg Medchemexpress, Rhodamine 110 CAS:13558-31-1 Purity:>98%
SDP
Catalog No. 501933186
Encompass_Preferred
Medchemexpress, HY-D0817 50mg Rhodamine 110 CAS:13558-31-1 Rhodamine 110 is a xanthene dye similar to fluorescein and eosin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3186 Supplier Medchemexpress LLC Supplier No. HYD081750MG
Medchemexpress, HY-D0817 50mg Rhodamine 110 CAS:13558-31-1 Rhodamine 110 is a xanthene dye similar to fluorescein and eosin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.