Learn More
CPC Scientific (Z-Ala-Arg)2-R110 (hydrochloride salt) 0.5MG
SDP
Catalog No. 502110458
Encompass
Typsin.SEQUENCE: (Z-Ala-Arg)2-R110 (hydrochloride salt)ONE-LETTER SEQUENCE: (Z-AR)2-R110MOLECULAR FORMULA: C54H60N12O11MOLECULAR WEIGHT: 1053.13STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: InflammationREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-002A, SUBS-002B, SUBS-002C, SUBS-002D
Catalog No. 50-211-0458 Supplier CPC Scientific Supplier No. SUBS002A
Typsin.SEQUENCE: (Z-Ala-Arg)2-R110 (hydrochloride salt)ONE-LETTER SEQUENCE: (Z-AR)2-R110MOLECULAR FORMULA: C54H60N12O11MOLECULAR WEIGHT: 1053.13STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: InflammationREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-002A, SUBS-002B, SUBS-002C, SUBS-002D