Learn More
CPC Scientific (Z-Ile-Pro-Arg)2-R110 (hydrochloride salt) 25MG
SDP
Catalog No. 502110467
Encompass
Typsin.SEQUENCE: (Z-Ile-Pro-Arg)2-R110 (hydrochloride salt)ONE-LETTER SEQUENCE: (Z-IPR)2-R110MOLECULAR FORMULA: C70H86N14O13MOLECULAR WEIGHT: 1331.52 (HCl salt not included in MW)STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: BZiPARRESEARCH AREA: InflammationREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-004A, SUBS-004B, SUBS-004C, SUBS-004D
Catalog No. 50-211-0467 Supplier CPC Scientific Supplier No. SUBS004D
Typsin.SEQUENCE: (Z-Ile-Pro-Arg)2-R110 (hydrochloride salt)ONE-LETTER SEQUENCE: (Z-IPR)2-R110MOLECULAR FORMULA: C70H86N14O13MOLECULAR WEIGHT: 1331.52 (HCl salt not included in MW)STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: BZiPARRESEARCH AREA: InflammationREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-004A, SUBS-004B, SUBS-004C, SUBS-004D