Learn More
CPC Scientific (Z-Phe-Arg)2-R110 (hydrochloride salt) 5MG
SDP
Catalog No. 502110470
Encompass
This item has a minimum order quantity of 3 per supplier requirements/
Plasmin, Cathepsin L.ONE-LETTER SEQUENCE: (Z-FR)2-R110MOLECULAR FORMULA: C66H68N12O11MOLECULAR WEIGHT:1205.32 (HCl salt not included in MW)STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Biol Chem Hoppe-Seyler 373, 433 (1992); Nature 361, 274 (1993)
Catalog No. 50-211-0470 Supplier CPC Scientific Supplier No. SUBS005C
This item has a minimum order quantity of 3 per supplier requirements/
Plasmin, Cathepsin L.ONE-LETTER SEQUENCE: (Z-FR)2-R110MOLECULAR FORMULA: C66H68N12O11MOLECULAR WEIGHT:1205.32 (HCl salt not included in MW)STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Biol Chem Hoppe-Seyler 373, 433 (1992); Nature 361, 274 (1993)