Learn More
CPC Scientific (H-Asp)2-R110 1MG
SDP
Catalog No. 502109036
Encompass
ONE-LETTER SEQUENCE: (D)2-R110MOLECULAR FORMULA: C28H25N4O9MOLECULAR WEIGHT:561.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Science 281, 1312 (1998); J. Biol. Chem., 276, 6974 (2001); Biochemistry 38, 13906 (1999)
Catalog No. 50-210-9036 Supplier CPC Scientific Supplier No. CASP065B
ONE-LETTER SEQUENCE: (D)2-R110MOLECULAR FORMULA: C28H25N4O9MOLECULAR WEIGHT:561.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Science 281, 1312 (1998); J. Biol. Chem., 276, 6974 (2001); Biochemistry 38, 13906 (1999)