Learn More
CPC Scientific (Tos-Gly-Pro-Arg)2-R110 (trifluoroacetate salt) 10MG
SDP
Catalog No. 502110475
Encompass
Thrombin.ONE-LETTER SEQUENCE: (Tos-GPR)2-R110MOLECULAR FORMULA: C60H70N14O13S2MOLECULAR WEIGHT:1259.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: HematologyREFERENCES: Biol Chem Hoppe-Seyler 373, 433 (1992); Nature 361, 274 (1993)
Catalog No. 50-211-0475 Supplier CPC Scientific Supplier No. SUBS006D
Thrombin.ONE-LETTER SEQUENCE: (Tos-GPR)2-R110MOLECULAR FORMULA: C60H70N14O13S2MOLECULAR WEIGHT:1259.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: HematologyREFERENCES: Biol Chem Hoppe-Seyler 373, 433 (1992); Nature 361, 274 (1993)