Learn More
CPC Scientific (Suc-Ala-Ala-Pro-Phe)2-R110 0.5MG
SDP
Catalog No. 502110476
Encompass
Chymotrypsin.SEQUENCE: (Suc-Ala-Ala-Pro-Phe)2-R110ONE-LETTER SEQUENCE: (Suc-AAPF)2-R110MOLECULAR FORMULA: C68N74N10O17MOLECULAR WEIGHT: 1304.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-007A, SUBS-007B
Catalog No. 50-211-0476 Supplier CPC Scientific Supplier No. SUBS007A
Chymotrypsin.SEQUENCE: (Suc-Ala-Ala-Pro-Phe)2-R110ONE-LETTER SEQUENCE: (Suc-AAPF)2-R110MOLECULAR FORMULA: C68N74N10O17MOLECULAR WEIGHT: 1304.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:Biol Chem Hoppe-Seyler 373, 433 (1992)Nature 361, 274 (1993)SKU(s): SUBS-007A, SUBS-007B