Learn More
Biotium RHODAMINE 110 BZIPAR 5mg
Catalog No. 501964122
Encompass_Preferred
BZiPAR (Rhodamine 110, bis-(N-CBZ-L-isoleucyl-L-prolyl-L-arginine amide), dihydrochloride) is a substrate for trypsin. The substrate has been reported to enter live cells and be hydrolyzed by lysosomal proteases.Properties: Ex/Em of end product (R110) = 496/520 nm; Off-white to pink solid soluble in DMSO; Store at 4°C and protect from light; C70 H88Cl2N14O13; MW: 1404.5
Catalog No. 50-196-4122 Supplier Biotium Supplier No. 10208
BZiPAR (Rhodamine 110, bis-(N-CBZ-L-isoleucyl-L-prolyl-L-arginine amide), dihydrochloride) is a substrate for trypsin. The substrate has been reported to enter live cells and be hydrolyzed by lysosomal proteases.Properties: Ex/Em of end product (R110) = 496/520 nm; Off-white to pink solid soluble in DMSO; Store at 4°C and protect from light; C70 H88Cl2N14O13; MW: 1404.5