Learn More
AdipoGen Rhodamine 110 chloride, 13558-31-1, MFCD00042009, 1 g
SDP
Catalog No. 501147231
Encompass_Preferred
MF: C20H14N2O3 . HCl, MW: 366.802, ≥97%, Appearance: Red to red-brown crystalline or powder. Fluorescent dye. Fluorophore standard that is widely used in enzyme assay systems. Precursor for a variety of R110-based enzyme substrates for flow cytometric measurements. Used as a calibration standard for R110-based enzyme substrates. Compared to AMC and AFC enzyme substrates, fluorogenic R110 substrates are generally more sensitive, giving longer absorption and fluorescence wavelength than AMC and AFC substrates.
Catalog No. 50-114-7231 Supplier AdipoGen Supplier No. CDXR0016G001
MF: C20H14N2O3 . HCl, MW: 366.802, ≥97%, Appearance: Red to red-brown crystalline or powder. Fluorescent dye. Fluorophore standard that is widely used in enzyme assay systems. Precursor for a variety of R110-based enzyme substrates for flow cytometric measurements. Used as a calibration standard for R110-based enzyme substrates. Compared to AMC and AFC enzyme substrates, fluorogenic R110 substrates are generally more sensitive, giving longer absorption and fluorescence wavelength than AMC and AFC substrates.