Learn More
AdipoGen Rhodamine 110 chloride,Chemical. CAS: 13558-31-1.
SDP
Catalog No. 501147232
Encompass_Preferred
Rhodamine 110 chloride, AdipoGen Life Sciences. Chemical. CAS: 13558-31-1. Formula: C20H14N2O3 . HCl. MW: 330.3 . 36.5. Synthetic Fluorescent dye. Fluorophore standard that is widely used in enzyme assay systems. Precursor for a variety of R110-based enzyme substrates for flow cytometric measurements. Used as a calibration standard for R110-based enzyme substrates. Compared to AMC and AFC enzyme substrates, fluorogenic R110 substrates are generally more sensitive, giving longer absorption and fluorescence wavelength than AMC and AFC substrates. Spectral data: lambdaex 498nm| lambdaem 520nm in methanol.
Catalog No. 50-114-7232 Supplier AdipoGen Supplier No. CDXR0016M250
Rhodamine 110 chloride, AdipoGen Life Sciences. Chemical. CAS: 13558-31-1. Formula: C20H14N2O3 . HCl. MW: 330.3 . 36.5. Synthetic Fluorescent dye. Fluorophore standard that is widely used in enzyme assay systems. Precursor for a variety of R110-based enzyme substrates for flow cytometric measurements. Used as a calibration standard for R110-based enzyme substrates. Compared to AMC and AFC enzyme substrates, fluorogenic R110 substrates are generally more sensitive, giving longer absorption and fluorescence wavelength than AMC and AFC substrates. Spectral data: lambdaex 498nm| lambdaem 520nm in methanol.