Learn More
Selleck Chemical LLC Rhodamine 123 S3577-25mg
SDP
Catalog No. 503109243
Encompass_Preferred
Rhodamine 123 (RH-123 R-22420) is a fluorescent cationic dye used to label mitochondria in living cells Rhodamine 123 inhibits ADP-stimulated respiration of mitochondria with Ki 12 M and ATPase activity of inverted inner membrane vesicles with Ki of 126 M and partially purified F1-ATPase with Ki of 177 M
Catalog No. 50-310-9243 Supplier Selleck Chemical LLC Supplier No. S357725MG
Rhodamine 123 (RH-123 R-22420) is a fluorescent cationic dye used to label mitochondria in living cells Rhodamine 123 inhibits ADP-stimulated respiration of mitochondria with Ki 12 M and ATPase activity of inverted inner membrane vesicles with Ki of 126 M and partially purified F1-ATPase with Ki of 177 M