Learn More
Abcam Rhodamine 123, 25MG
Catalog No. 50996121
Encompass_Preferred
MW 380.8 Da, Purity >98%. Cell-permeable, green fluorescent dye suitable for monitoring membrane polarization in mitochondria and bacteria in live cell assays. Accumulation in membranes is dependent on membrane polarization. Also used in fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA. Absorption peak at 505 nm. Luminescence tunable at ~560 nm when used as a laser dye.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-121 Supplier Abcam Supplier No. AB27554525MG
MW 380.8 Da, Purity >98%. Cell-permeable, green fluorescent dye suitable for monitoring membrane polarization in mitochondria and bacteria in live cell assays. Accumulation in membranes is dependent on membrane polarization. Also used in fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA. Absorption peak at 505 nm. Luminescence tunable at ~560 nm when used as a laser dye.
The product is subject to the following: Abcam Restricted Use Statement