Learn More
Biotium Rhodamine 123
Catalog No. 501964745
Encompass_Preferred
Rhodamine 123 is a popular green fluorescent mitochondrial dye that stains mitochondria in living cells in a membrane potential-dependent fashion. It is widely used in flow cytometry studies involving mitochondrial membrane potential. Please also see Dihydrorhodamine 123 and its HCl salt.Properties: Ex/Em(MeOH) = 505/534 nm; e (505 nm, MeOH) = 97,000; Orange red solid soluble in DMSO or DMF; Store at 4°C and protect from light, especially in solution; C21H17ClN2O3; MW: 381; [62669-70-9]
Catalog No. 50-196-4745 Supplier Biotium Supplier No. 70010
Rhodamine 123 is a popular green fluorescent mitochondrial dye that stains mitochondria in living cells in a membrane potential-dependent fashion. It is widely used in flow cytometry studies involving mitochondrial membrane potential. Please also see Dihydrorhodamine 123 and its HCl salt.Properties: Ex/Em(MeOH) = 505/534 nm; e (505 nm, MeOH) = 97,000; Orange red solid soluble in DMSO or DMF; Store at 4°C and protect from light, especially in solution; C21H17ClN2O3; MW: 381; [62669-70-9]