Learn More
TARGETMOL CHEMICALS INC Rhodamine 123 50MG
SDP
Catalog No. 502879636
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Rhodamine 123 is a fluorescent dye (λex=503 nm, λem=527 nm). Purity 99.23%
Catalog No. 50-287-9636 Supplier TARGETMOL CHEMICALS INC Supplier No. T1674350MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Rhodamine 123 is a fluorescent dye (λex=503 nm, λem=527 nm). Purity 99.23%