Learn More
Medchemexpress LLC HY-D0816 5mg Medchemexpress, Rhodamine 123 CAS:62669-70-9 Purity:>98%
SDP
Catalog No. 501933185
Encompass_Preferred
Medchemexpress, HY-D0816 5mg Rhodamine 123 CAS:62669-70-9 Rhodamine 123 (RH-123; R-22420) is a fluorescent dye (λ ex =503 nm, λ em =527 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3185 Supplier Medchemexpress LLC Supplier No. HYD08165MG
Medchemexpress, HY-D0816 5mg Rhodamine 123 CAS:62669-70-9 Rhodamine 123 (RH-123; R-22420) is a fluorescent dye (λ ex =503 nm, λ em =527 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.