Learn More
Medchemexpress LLC Rhodamine 123 | 62669-70-9 | 99.5% | 1 ML
SDP
Catalog No. 5000410936
Encompass_Preferred
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials. These probes attach to mitochondria and produce bright fluorescence. At certain concentrations, rhodamine dyes have low toxicity to cells, making them commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Recognizes mitochondrial membrane potentials
  • Used for cell staining analysis
  • Suitable for detecting mitochondria in various cell types
  • Low toxicity at specific concentrations
Catalog No. 50-004-10936 Supplier Medchemexpress LLC Supplier No. HYD081610MM/1ML
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials. These probes attach to mitochondria and produce bright fluorescence. At certain concentrations, rhodamine dyes have low toxicity to cells, making them commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Recognizes mitochondrial membrane potentials
  • Used for cell staining analysis
  • Suitable for detecting mitochondria in various cell types
  • Low toxicity at specific concentrations