Learn More
Medchemexpress LLC Rhodamine 123 | 62669-70-9 | 99.5% | 100 MG
SDP
Catalog No. 5000410280
Encompass_Preferred
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials, thereby attaching to mitochondria and producing bright fluorescence. At certain concentrations, rhodamine dyes have low toxicity to cells, making them commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Specifically recognizes mitochondrial membrane potentials
  • Attaches to mitochondria and produces bright fluorescence
  • Low toxicity to cells at certain concentrations
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms
Catalog No. 50-004-10280 Supplier Medchemexpress LLC Supplier No. HYD0816100MG
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials, thereby attaching to mitochondria and producing bright fluorescence. At certain concentrations, rhodamine dyes have low toxicity to cells, making them commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Specifically recognizes mitochondrial membrane potentials
  • Attaches to mitochondria and produces bright fluorescence
  • Low toxicity to cells at certain concentrations
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms