Learn More
Medchemexpress LLC Rhodamine 123 | 62669-70-9 | 99.5% | 50 MG
SDP
Catalog No. 5000410937
Encompass_Preferred
Rhodamine 123 is a membrane-permeable cationic fluorescent probe that specifically recognizes mitochondrial membrane potentials. It attaches to mitochondria, producing bright fluorescence. At certain concentrations, rhodamine dyes exhibit low toxicity to cells, making them commonly used for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe.
  • Specifically recognizes mitochondrial membrane potentials.
  • Attaches to mitochondria and produces bright fluorescence.
  • Low toxicity to cells at certain concentrations.
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.
Catalog No. 50-004-10937 Supplier Medchemexpress LLC Supplier No. HYD081650MG
Rhodamine 123 is a membrane-permeable cationic fluorescent probe that specifically recognizes mitochondrial membrane potentials. It attaches to mitochondria, producing bright fluorescence. At certain concentrations, rhodamine dyes exhibit low toxicity to cells, making them commonly used for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe.
  • Specifically recognizes mitochondrial membrane potentials.
  • Attaches to mitochondria and produces bright fluorescence.
  • Low toxicity to cells at certain concentrations.
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms.